Learn More
CARS Antibody (CL2310) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-36758MFV500
Product Code. 30290695
  • £574.00 / 0.10mL
Description
CARS Monoclonal antibody specifically detects CARS in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CARS
Monoclonal
mFluor Violet 500 SE
CARS1, CysRS, cysteine translase, cysteine tRNA ligase 1, cytoplasmic, Cysteine--tRNA ligase, cysteinyl-tRNA synthetase, cysteinyl-tRNA synthetase, cytoplasmic, EC 6.1.1, EC 6.1.1.16, MGC:11246
This antibody was developed using a recombinant protein derived from A8MVQ3, with the exact immunogen sequence remaining proprietary.
0.1 mL
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
833
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL2310
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified