missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARS Antibody (CL2309), Novus Biologicals™
Mouse Monoclonal Antibody
£222.00 - £470.00
Specifications
| Antigen | CARS |
|---|---|
| Clone | CL2309 |
| Dilution | Immunohistochemistry 1:2500-1:5000 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626605
|
Novus Biologicals
NBP2-36757-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18192249
|
Novus Biologicals
NBP2-36757 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CARS Monoclonal specifically detects CARS in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CARS | |
| Immunohistochemistry 1:2500-1:5000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CARS1, CysRS, cysteine translase, cysteine tRNA ligase 1, cytoplasmic, Cysteine--tRNA ligase, cysteinyl-tRNA synthetase, cysteinyl-tRNA synthetase, cytoplasmic, EC 6.1.1, EC 6.1.1.16, MGC:11246 | |
| CARS | |
| IgG2b | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL2309 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| A8MVQ3 | |
| 833 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SQCNLYMAARKAVRKRPNQALLENIALYLTHMLKIFGAVEEDSSLGFPVGGPGTSLSLEATVMPYLQVLSEFREGVRKIAREQKVPEILQLSDALRDNILPELGVRFEDHEGLPTVVKLVDRNTLLKEREEKRRVE | |
| Primary | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title