missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARS Antibody (CL2309) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36757AF700
This item is not returnable.
View return policy
Description
CARS Monoclonal antibody specifically detects CARS in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| CARS | |
| Monoclonal | |
| Alexa Fluor 700 | |
| CARS1, CysRS, cysteine translase, cysteine tRNA ligase 1, cytoplasmic, Cysteine--tRNA ligase, cysteinyl-tRNA synthetase, cysteinyl-tRNA synthetase, cytoplasmic, EC 6.1.1, EC 6.1.1.16, MGC:11246 | |
| This antibody was developed using a recombinant protein derived from A8MVQ3, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 833 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL2309 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction