missing translation for 'onlineSavingsMsg'
Learn More

Whirl-pak™ Carrying Rack - 18 Compartment
GREENER_CHOICE

Product Code. 16861590 Shop All Whirl-Pak Products
Change view
Click to view available options
Material:
Steel
Unit Size:
Each
Product Code. Material unitSize
16861590 Steel Each
1 options
This item is not returnable. View return policy

Product Code. 16861590

Brand: Whirl-pak™ B00772WA

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specially designed to hold Whirl-Pak™ Bags

  • Carrying rack with 18 compartments
  • Specially designed to hold Whirl-Pak™ Bags
  • 10 and 12-gauge steel is fused together to create strong joints and straight lines
  • Rack is finished with an electrostatically applied powder coating
  • Strong, durable finish which will stand up to many laboratory materials
  • Fits into regular-size Playmate™ Coolers and Nasco™ Milk Sample Ice Chests
  • Autoclavable
  • Made in the USA

Specifications

Autoclavable Autoclavable
Color Blue
Material Steel
Positions 18 Compartments
Sterility Sterile
For Use With (Equipment) Whirl-Pak™ Bags
Quantity 1 Each
Dimensions (L x W) 2 x 1.625 in. (Compartment)
Dimensions (L x W x H) 10.75 x 6.5 x 3 in. (27.3 x 16.5 x 7.6 cm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.