missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Carrying Kit Set for Eutech™ ph 5+ and ph 6+ meters

Product Code. 12973507
This item is not returnable. View return policy
Product Code. 12973507 Supplier Thermo Scientific™ Supplier No. 01X266901
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Eutech™ Carrying Kit Cases are thoughtfully designed to hold your Eutech™ meter and electrodes.

Made of sturdy reinforced plastic, the kit comes complete with calibrating sachets and sample bottles, and is thoughtfully designed to hold your meter and electrodes. It's all you need for your outfield measurements. The Eutech™ Economy Carrying Kit Set is used for Eutech™ pH 5+ and pH 6+ meters.

The Carrying Kit set is comprised of:

  • pH 4.01 Buffer Solution
  • pH 7.00 Buffer Solution
  • Storage Solution
  • Deionised rinse water (60 ml)
  • TRUSTED_SUSTAINABILITY

    Specifications

    Description Carrying Kit Set for Eutech™ ph 5+ and ph 6+ meters
    Type Accessory
    Includes pH 4.01 Buffer Solution, pH 7.00 Buffer Solution, Storage Solution, Deionised Rinse Water (60 ml Each)
    For Use With (Equipment) Eutech™ ph 5+ and ph 6+ meters
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.