Learn More
Melford Carrageenan Kappa
Description
Kappa carrageenan binds water to form strong, rigid gels in the presence of potassium salts.
As the level of potassium is increased, the gel structure becomes tightly aggregated and may result in syneresis (moisture on the gel surface).
Specifications
Specifications
| Quantity | 100 g |