missing translation for 'onlineSavingsMsg'
Learn More

Carpipramine Dihydrochloride, TRC

Product Code. 30005539
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005539 1 mg 1mg
30012652 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005539 Supplier Toronto Research Chemicals Supplier No. TRCC1843201MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7075-03-8
Molecular Formula C28 H37 Cl N4 O . 2 Cl H
Molecular Weight (g/mol) 553.99
Synonym [1,4'-Bipiperidine]-4'-carboxamide, 1'-[3-(10,11-dihydro-5H-dibenz[b,f]azepin-5-yl)propyl]-, hydrochloride (1:2), [1,4'-Bipiperidine]-4'-carboxamide, 1'-[3-(10,11-dihydro-5H-dibenz[b,f]azepin-5-yl)propyl]-, dihydrochloride (7CI,8CI,9CI), 5H-Dibenz[b,f]azepine, [1,4'-bipiperidine]-4'-carboxamide deriv., Carpipramine dihydrochloride, Defecton, Defekton, PZ 1511
IUPAC Name 1-[3-(2-chloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)propyl]-4-piperidin-1-ylpiperidine-4-carboxamide;dihydrochloride
SMILES Cl.Cl.NC(=O)C1(CCN(CCCN2c3ccccc3CCc4ccc(Cl)cc24)CC1)N5CCCCC5

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C28H37ClN4O.2ClH/c29-24-12-11-23-10-9-22-7-2-3-8-25(22)33(26(23)21-24)18-6-15-31-19-13-28(14-20-31,27(30)34)32-16-4-1-5-17-32;;/h2-3,7-8,11-12,21H,1,4-6,9-10,13-20H2,(H2,30,34);2*1H
Formula Weight 552.2189
Chemical Name or Material Carpipramine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.