missing translation for 'onlineSavingsMsg'
Learn More

Carphenazine, TRC

Product Code. 17962535
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962535 50 mg 50mg
17954145 25 mg 25mg
17935105 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17962535 Supplier Toronto Research Chemicals Supplier No. TRCC18430550MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2622-30-2
Molecular Formula C24 H31 N3 O2 S
Molecular Weight (g/mol) 425.59
Synonym 1-[10-[3-[4-(2-Hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-1-propanone Carfenazine, 1-Propanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl]- (6CI,7CI,8CI), Carfenazine, Carphenazin, Carphenazine, Proketazine
IUPAC Name 1-[10-[3-[4-(2-hydroxyethyl)piperazin-1-yl]propyl]phenothiazin-2-yl]propan-1-one
SMILES CCC(=O)c1ccc2Sc3ccccc3N(CCCN4CCN(CCO)CC4)c2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H31N3O2S/c1-2-22(29)19-8-9-24-21(18-19)27(20-6-3-4-7-23(20)30-24)11-5-10-25-12-14-26(15-13-25)16-17-28/h3-4,6-9,18,28H,2,5,10-17H2,1H3
Formula Weight 425.2137
Chemical Name or Material Carphenazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.