missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CARP Recombinant Rabbit Monoclonal Antibody (2S10L8)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555387
This item is not returnable.
View return policy
Description
Immunogen Sequence: MMVLKVEELV TGKKNGNGEA GEFLPEDFRD GEYEAAVTLE KQEDLKTLLA HPVTLGEQQW KSEKQREAEL KKKKLEQRSK LENLEDLEII IQLKKRKKYR KTKVPVVKEP EPEIITEPVD VPTFLKAALE.
The protein encoded by this gene is localized to the nucleus of endothelial cells and is induced by IL-1 and TNF-alpha stimulation. Studies in rat cardiomyocytes suggest that this gene functions as a transcription factor. Interactions between this protein and the sarcomeric proteins myopalladin and titin suggest that it may also be involved in the myofibrillar stretch-sensor system.
Specifications
| CARP | |
| Recombinant Monoclonal | |
| 0.87 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q15327, Q8R560, Q9CR42 | |
| Ankrd1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human ANKRD1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 2S10L8 | |
| Unconjugated | |
| Ankrd1 | |
| ALRP; ANKRD1; ankyrin repeat domain 1; ankyrin repeat domain 1 (cardiac muscle); ankyrin repeat domain-containing protein 1; ankyrin-like repeat protein; bA320F15.2; C193; C-193; Cardiac adriamycin-responsive protein; cardiac ankyrin repeat protein; cardiac responsive adriamycin protein; Carp; Crap; CVARP; Cytokine-inducible gene C-193 protein; cytokine-inducible nuclear protein; HA1A2; liver ankyrin repeat domain 1; MARP; MARP1; MCARP; muscle ankyrin repeat protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 107765, 27063, 27064 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction