missing translation for 'onlineSavingsMsg'
Learn More

β-Carotene, MedChemExpress

Product Code. p-200101484
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16484827

Brand: Medchem Express HYN0411

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress β-Carotene (Provitamin A), a carotenoid compound, is a naturally-occurring vitamin A precursor.

β-Carotene is a modulator of reactive oxygen species (ROS), with antioxidant and antiinflammatory activities. β-Carotene may serve as an antioxidant or as a prooxidant, depending on its intrinsic properties as well as on the redox potential of the biological environment in which it acts. β-Carotene induces breast cancer cells apoptosis, with anticancer activities.

Specifications

Chemical Name or Material β-Carotene
CAS 7235-40-7
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C40H56
Quantity 50 mg
Synonym Provitamin A beta-Carotene
Solubility Information THF : 20 mg/mL (37.25 mM; Need ultrasonic) ∣Acetone : 5 mg/mL (9.31 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.86 mM; ultrasonic and warming and heat to 60°C)
SMILES CC1(C)C(/C=C/C(C)=C/C=C/C(C)=C/C=C/C=C(C)/C=C/C=C(C)/C=C/C2=C(C)CCCC2(C)C)=C(C)CCC1
Molecular Weight (g/mol) 536.87
Formula Weight 536.87
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.