missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carnosine Dipeptidase 1/CNDP1 Antibody (CL0339), Novus Biologicals™
Mouse Monoclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | Carnosine Dipeptidase 1/CNDP1 |
|---|---|
| Clone | CL0339 |
| Dilution | Western Blot 1 ug/ml |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18435282
|
Novus Biologicals
NBP2-30972-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18126262
|
Novus Biologicals
NBP2-30972 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carnosine Dipeptidase 1/CNDP1 Monoclonal specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for Western Blot.Specifications
| Carnosine Dipeptidase 1/CNDP1 | |
| Western Blot 1 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase | |
| CNDP1 | |
| IgG1 | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL0339 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| Q96KN2 | |
| 84735 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PALLEKVFQYIDLHQDEFVQTLKEWVAIESDSVQPVPRFRQELFRMMAVAADTLQRLGARVASVDMGPQQLPDGQSLPIPPVILAELGSDPTKGTVCFYGHL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title