missing translation for 'onlineSavingsMsg'
Learn More

Carnosine Dipeptidase 1/CNDP1 Antibody (352216) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30732796 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732796 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732796 Supplier Novus Biologicals Supplier No. FAB2489I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Carnosine Dipeptidase 1/CNDP1 Monoclonal antibody specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carnosine Dipeptidase 1/CNDP1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 352216
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Carnosine Dipeptidase 1/CNDP1, Pro28-His507, Accession # Q96KN2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84735
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.