missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carnosine Dipeptidase 1/CNDP1 Antibody (001), Janelia Fluor™ 549, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Carnosine Dipeptidase 1/CNDP1 Monoclonal specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Carnosine Dipeptidase 1/CNDP1 |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1 |
| Conjugate | Janelia Fluor 549 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Carnosine Dipeptidase 1/CNDP1 (NP_116038.4; Ser 27-His 508). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?