missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carnosine Dipeptidase 1/CNDP1 Antibody (001), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89263CL1
This item is not returnable.
View return policy
Description
Carnosine Dipeptidase 1/CNDP1 Monoclonal antibody specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for ELISA,Immunocytochemistry/Immunofluorescence
Specifications
| Carnosine Dipeptidase 1/CNDP1 | |
| Monoclonal | |
| CoraFluor 1 | |
| beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carnosine Dipeptidase 1/CNDP1 (Accession#: NP_116038.4; Ser27-His507). | |
| 0.1 mL | |
| Signal Transduction | |
| 84735 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| 001 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction