missing translation for 'onlineSavingsMsg'
Learn More

(±)-Carnitine chloride, MedChemExpress

Product Code. 16441118
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441118 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441118 Supplier Medchem Express Supplier No. HYB1453
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (±)-Carnitine chloride exists in two isomers, known as D and L. L-carnitine plays an essential role in the β-oxidation of fatty acids and also shows antioxidant, and anti-inflammatory activities.

(±)-Carnitine chloride exists in two isomers, known as D and L. L-carnitine plays an essential role in the β-oxidation of fatty acids and also shows antioxidant, and anti-inflammatory activities.

Specifications

Chemical Name or Material (±)-Carnitine chloride
CAS 461-05-2
Color White
Purity Grade Notes Research
Molecular Formula C7H16ClNO3
Quantity 5 g
Synonym DL-Carnitine chloride
Solubility Information H2O : ≥ 100 mg/mL (505.92 mM) ∣DMSO : 25 mg/mL (126.48 mM; Need ultrasonic)
SMILES C[N+](C)(C)CC(O)CC(O)=O.[Cl-]
Molecular Weight (g/mol) 197.66
Formula Weight 197.66
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.