missing translation for 'onlineSavingsMsg'
Learn More

Carnauba wax

Product Code. 11337408
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337408 250 g 250g
11327408 1 kg 1kg
11317408 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11337408 Supplier Thermo Scientific Alfa Aesar Supplier No. 031117.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8015-86-9

Carnauba wax is used as a glazing agent, bulking agent, acidity regulator and tablet-coating agent. It is also used in automobile waxes, shoe polishes, dental floss and food products such as sweets, instrument polishes, floor, furniture waxes, polishes and paper coatings. It serves as the main ingredient in surfboard wax. Further, it is also used as a mold release agent to produce fiber-reinforced plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8015-86-9
MDL Number MFCD00130724
Synonym Wax

Specifications

Melting Point 82°C to 86°C
Density 0.990 to 0.999
Odor Odorless
Refractive Index 1.45
Quantity 250 g
Merck Index 14,1846
Solubility Information Soluble in diethyl ether,alkali,chloroform. Slightly soluble in boiling ethanol. Insoluble in water.
Physical Form Flakes
Chemical Name or Material Carnauba wax

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.