missing translation for 'onlineSavingsMsg'
Learn More

Carmofur, MedChemExpress

Product Code. 16442056
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16442056

Brand: Medchem Express HYB0182500mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Carmofur (HCFU), a derivative of 5-Fluorouracil, is an antineoplastic agent. Carmofur is an inhibitor of acid ceramidase with an IC50 of 79 nM for the rat enzyme. Carmofur inhibits the SARS-CoV-2 main protease (Mpro). Carmofur Inhibits SARS-CoV-2 in Vero E6 cell with an EC50 of 24.3 μM.

Carmofur (HCFU), a derivative of 5-Fluorouracil, is an antineoplastic agent. Carmofur is an inhibitor of acid ceramidase with an IC50 of 79 nM for the rat enzyme. Carmofur inhibits the SARS-CoV-2 main protease (Mpro). Carmofur Inhibits SARS-CoV-2 in Vero E6 cell with an EC50 of 24.3 μM.

Specifications

Chemical Name or Material Carmofur
Physical Form Solid
CAS 61422-45-5
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C11H16FN3O3
For Use With (Application) Cancer-programmed cell death
Synonym HCFU
Solubility Information DMSO : ≥ 100 mg/mL (388.71 mM) ∣H2O : 0.67 mg/mL (2.60 mM; Need ultrasonic)
SMILES O=C(N(C(N1)=O)C=C(F)C1=O)NCCCCCC
Molecular Weight (g/mol) 257.26
Formula Weight 257.26
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.