missing translation for 'onlineSavingsMsg'
Learn More

Carmine (C.I. 75470) (Technical Grade), TRC

Product Code. 30135331
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135331 50 mg 50mg
30007791 100 mg 100mg
30003671 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135331 Supplier Toronto Research Chemicals Supplier No. TRCC18365350MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1390-65-4
Molecular Formula C22H15AlCaO13
Molecular Weight (g/mol) 554.41
Synonym B Rose Liquid, CC 1000WS, Carmine red
IUPAC Name aluminum;calcium;1-methyl-3,5,6,8-tetraoxido-9,10-dioxo-7-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylate
SMILES [Al+3].[Ca+2].Cc1c(C(=O)[O-])c([O-])cc2C(=O)c3c([O-])c([O-])c([C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c([O-])c3C(=O)c12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H20O13.Al.Ca/c1-4-8-5(2-6(24)9(4)22(33)34)13(25)10-11(15(8)27)16(28)12(18(30)17(10)29)21-20(32)19(31)14(26)7(3-23)35-21;;/h2,7,14,19-21,23-24,26,28-32H,3H2,1H3,(H,33,34);;/q;+3;+2/p-5/t7-,14-,19+,20-,21+;;/m1../s1
Formula Weight 553.9954
Chemical Name or Material Carmine (C.I. 75470) (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.