missing translation for 'onlineSavingsMsg'
Learn More

Carm1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18355795 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355795 20 μg 20µL
18398631 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18355795 Supplier Novus Biologicals Supplier No. NBP31689220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Carm1 Polyclonal antibody specifically detects Carm1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carm1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias coactivator-associated arginine methyltransferase 1EC 2.1.1.-, EC 2.1.1, histone-arginine methyltransferase CARM1, PRMT4EC 2.1.1.125, Protein arginine N-methyltransferase 4
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around S228 of human PRMT4/Carm1 (NP_954592.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 10498
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.