missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carm1 Antibody (CARM1/7426), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20591-100ug
This item is not returnable.
View return policy
Description
Carm1 Monoclonal antibody specifically detects Carm1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Carm1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| coactivator-associated arginine methyltransferase 1EC 2.1.1.-, EC 2.1.1, histone-arginine methyltransferase CARM1, PRMT4EC 2.1.1.125, Protein arginine N-methyltransferase 4 | |
| Recombinant fragment (around aa200-400) of human Carm1 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Cellular Markers, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 10498 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CARM1/7426 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction