Learn More
Medline Scientific™ Planachromatic Objective
Description
- For use with Magnum series biological microscopes or Inverso TC EPI microscopes
- Creates a full, flat field image, ideal for imaging purposes
- 5x / 0.12 / 160 / 0.17 or 4x / 0.10 / 160 / 1.2 LWD
Specifications
Specifications
| Type | Objective |
| For Use With (Equipment) | Inverso TC EPI microscopes |
| Magnification Power | 4X |
| Quantity | 1 Ea. |