Learn More
Motic™ CCIS™ Plan Achromatic Phase Objective
Product Code. 11752348
Click to view available options
Type:
PL Ph0 4X
PL Ph1 10X
Focus Distance:
0.1 mm, WD=12.6 mm
0.25 mm, WD=4.1 mm
Unit Size:
Each
Product Code. 11752348
Brand: Motic™ 1101001703281
Only null left
Add to basket
Add to basket
Provides a flat field of view while also correcting for color aberration. Motic™ CCIS™ Plan Achromatic Phase Objective is useful on specimens that do not absorb light.
  • For use with AE31 Inverted Microscope
  • Achromatic Plan Phase Objective
  • Flat field
  • Eliminates color aberration
  • Magnification and Numerical Apertures available:
    • 10X/0.25 (WD=4.1mm) Ph1 for AE
    • 4X/0.10 (WD=12.6mm) Ph0 for AE
Specifications
Type PL Ph0 4X
Focus Distance 0.1 mm, WD=12.6 mm
For Use With (Equipment) AE31 inverted microscope
Quantity 1 Piece