Learn More
Motic™ Micrometer Eyepiece Reticle
Product Code. 11732298
Change view
Click to view available options
Type:
Reticle Eyepiece Micrometer
Unit Size:
Each
Product Code. Type Unit Size
11732298 Reticle Eyepiece Micrometer Each
1 options
Product Code. 11732298 Supplier Motic™ Supplier No. 1101001401841
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
Use with 10X eyepiece for BA Series Microscope. Motic™ Micrometer Eyepiece Reticle includes 100 divisions in 10mm
  • 100 divisions in 10mm
  • 25mm dia.
  • For use with BA series microscope 10X eyepiece
Specifications
Type Reticle Eyepiece Micrometer
Description 100 divisions in 10mm
Diameter (Metric) 25 mm
For Use With (Equipment) BA Series Microscope
Quantity 1 Piece