missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ LED Module 4500°K +/-300°K
Click to view available options
Color:
Bright White
Unit Size:
Each
Description
- LED module for use with inverted AE2000 or AE31 Elite Microscopes
- 4500K color temperature produces bright white task lighting
- Replaces halogen bulb
Specifications
Specifications
| Type | LED Module |
| Color | Bright White |
| Description | 4500°K color temperature |
| For Use With (Application) | Microscope illumination |
| Quantity | 1 Piece |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction