Learn More
Carl Zeiss™ Immersol™ Immersion Oil, 518 F; 20mL oiler
Brand:  Zeiss 444960-0000-000
Product Code. 12737327
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Increases the resolving power of a microscope. Zeiss Immersol™ 518 F, Immersion Oil is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.