Learn More
Invitrogen™ CARKL Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5101269
Product Code. 16687744
  • £448.00 / 100µL
Description
Antibody detects endogenous levels of total CARKL.
The protein encoded by this gene has weak homology to several carbohydrate kinases, a class of proteins involved in the phosphorylation of sugars as they enter a cell, inhibiting return across the cell membrane. Sequence variation between this novel gene and known carbohydrate kinases suggests the possibility of a different substrate, cofactor or changes in kinetic properties distinguishing it from other carbohydrate kinases. The gene resides in a region commonly deleted in cystinosis patients, suggesting a role as a modifier for the cystinosis phenotype. The genomic region is also rich in Alu repetitive sequences, frequently involved in chromosomal rearrangements.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CARKL
Polyclonal
Unconjugated
SHPK
4930431K22Rik; AI194947; AW260459; Carbohydrate kinase-like protein; CARKL; Sedoheptulokinase; SHK; Shpk
Rabbit
Affinity Chromatography
RUO
23729, 287479, 74637
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9D5J6, Q9UHJ6
SHPK
A synthesized peptide derived from human SHPK(Accession Q9UHJ6), corresponding to amino acid residues A33-S83.
100 μL
Primary
Human, Mouse, Rat