missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Carisoprodol-d7 (iso-propyl-d7), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Carisoprodol-D7 (isopropyl-D7) |
| CAS | 1218911-01-3 |
| Molecular Formula | C12 D7 H17 N2 O4 |
| Quantity | 0.005 g |
| Synonym | Carbamic acid, N-[1-(methyl-d3)ethyl-1,2,2,2-d4]-, 2-[[(aminocarbonyl)oxy]methyl]-2-methylpentyl ester (ACI), 2-[[(Aminocarbonyl)oxy]methyl]-2-methylpentyl N-[1-(methyl-d3)ethyl-1,2,2,2-d4]carbamate (ACI), Carisoprodol-d7, Carisoprodol-D7 (isopropyl-D7), 2-[(Carbamoyloxy)methyl]-2-methylpentyl isopropyl-d7-carbamate, (±)-Carisoprodol-d7 (iso-propyl-d7), (±)-Carisoprodol-d7 (iso-propyl-d7) |
| InChI Formula | InChI=1 S/C12H24N2O4/c1-5-6-12(4,7-17-10(13)15)8-18-11(16)14-9(2)3/h9H,5-8H2,1-4H3,(H2,13,15)(H,14,16)/i2D3,3D3,9 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(NC(=O)OCC(C)(CCC)COC(=O)N)C([2 H])([2 H])[2 H] |
| IUPAC Name | [2-(carbamoyloxymethyl)-2-methylpentyl] N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)carbamate |
| Molecular Weight (g/mol) | 267.3731 |
| Formula Weight | 267.2175 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction