missing translation for 'onlineSavingsMsg'
Learn More

Cariprazine hydrochloride, MedChemExpress

Product Code. 16456158
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456158 10 mg 10mg
16432297 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16456158 Supplier Medchem Express Supplier No. HY14763A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cariprazine hydrochloride is a novel antipsychotic drug candidate that exhibits high affinity for the D3 (Ki=0.085 nM) and D2 (Ki=0.49 nM) receptors, and moderate affinity for the 5-HT1A receptor (Ki=2.6 nM).

Cariprazine hydrochloride is a novel antipsychotic drug candidate that exhibits high affinity for the D3 (Ki=0.085 nM) and D2 (Ki=0.49 nM) receptors, and moderate affinity for the 5-HT1A receptor (Ki=2.6 nM).

Specifications

Chemical Name or Material Cariprazine hydrochloride
Physical Form Solid
CAS 1083076-69-0
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H33Cl3N4O
For Use With (Application) Neuroscience-Neuromodulation
Synonym RGH188 hydrochloride
Solubility Information DMSO : 6.67 mg/mL (14.38 mM; Need ultrasonic) ∣H2O : 2.86 mg/mL (6.17 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(N[C@H]1CC[C@H](CCN2CCN(C3=CC=CC(Cl)=C3Cl)CC2)CC1)N(C)C.[H]Cl
Molecular Weight (g/mol) 463.87
Formula Weight 463.87
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.