missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Carfilzomib

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Carfilzomib is a potent irreversible proteasome inhibitor. Preferentially inhibits the chymotrypsin-like β5 subunit of the constitutive 20S proteasome (IC50 = 5.2 nM) and the β5i subunit of the immunoproteasome 20Si (LMP7; IC50 = 14 nM) in vitro with minimal cross-reactivity to other proteases. Exhibits little or no effect on PGPH and T-L activities. Activates prosurvival autophagy and induces cell apoptosis. Acts synergistically with dexamethasone (Cat. No. 1126). Suppresses tumor growth in an in vivo xenograft model. Decreases bone resorption and enhances bone formation in non-tumor bearing mice.
Specifications
Specifications
| Chemical Name or Material | (αS)-α-[[2-(4-Morpholinyl)acetyl]amino]benzenebutanoyl-L-leucyl-N-[(1 S)-3-methyl-1-[[(2 R)-2-methyl-2-oxiranyl]carbonyl]butyl]-L-phenylalaninamide |
| CAS | 868540-17-4 |
| Quantity | 10 mg |
| Target | Proteasome Inhibitors |
| Molecular Formula | C40H57N5O7 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction