missing translation for 'onlineSavingsMsg'
Learn More

Carfentrazone (free acid) 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30310714
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310714 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310714 Supplier Dr. Ehrenstorfer Supplier No. DREA11043100ME100
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carfentrazone (free acid)
CAS 128621-72-7
Analyte or Component Names Carfentrazone (free acid)
Shipping Condition Room Temperature
Molecular Formula C13 H10 Cl2 F3 N3 O3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym α,2-Dichloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1H-1,2,4-triazol-1-yl]-4-fluorobenzenepropanoic acid, 2-Chloro-3-[2-chloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1H-1,2,4-triazol-1-yl]-4-fluorophenyl]propionic acid, Aim EC, Carfentrazone, Oratio
InChI Formula InChI=1S/C13H10Cl2F3N3O3/c1-5-19-21(13(24)20(5)12(17)18)10-3-6(2-8(15)11(22)23)7(14)4-9(10)16/h3-4,8,12H,2H2,1H3,(H,22,23)
SMILES CC1=NN(C(=O)N1C(F)F)c2cc(CC(Cl)C(=O)O)c(Cl)cc2F
IUPAC Name 2-chloro-3-[2-chloro-5-[4-(difluoromethyl)-3-methyl-5-oxo-1,2,4-triazol-1-yl]-4-fluorophenyl]propanoic acid
Solvent or Matrix Methanol
Molecular Weight (g/mol) 384.14
Quantity 1 mL
Formula Weight 383.0051 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.