missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CARF/CDKN2AIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30557
This item is not returnable.
View return policy
Description
CARF/CDKN2AIP Polyclonal antibody specifically detects CARF/CDKN2AIP in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| CARF/CDKN2AIP | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55602 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| CARFCDKN2A-interacting protein, CDKN2A interacting protein, collaborates/cooperates with ARF (alternate reading frame) protein, Collaborator of ARF, FLJ20036 | |
| Between 1 and 50 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido