missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cardiotin Monoclonal Antibody (SR-4)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA106310

Product Code. 11538792

  • £454.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA1-06310 detects cardiotin in human and porcine samples. MA1-06310 has sucessfully been used in Western blotting and immunohistochemistry. By Western blotting, it detects a ~300 kDa protein representing cardiotin from. The MA1-06310 immunogen is cardiotin purified from mouse spleen cells.

Cardiotin is a recently discovered, high molecular weight protein complex (~300 kDa) located in the longitudinal SR of cardiomyocytes. The cardiotin complex exists as 60 kDa and 100 kDa subunits. Both subunits contain the same amino-terminal 14 amino-acid sequence, and show high homology to human skeletal muscle alpha-actin. The tetrameric configuration of the cardiotin protein complex suggests that it exists as a transmembrane complex with the N-terminus at the cytoplasmic side of the membrane, able to interact with actin. Immunofluorescence studies colocalize cardiotin with SERCA 2a ATPase in the longitudinal sarcoplasmic reticulum (SR), although cardiotin is not expressed in junctional SR with SERCA 2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cardiotin
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
Cardiotin purified from mouse spleen cells.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Frozen), Western Blot
SR-4
Unconjugated
Mouse
Purified
RUO
Human, Pig
Antibody
IgM
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.