missing translation for 'onlineSavingsMsg'
Learn More

Cardiogenol C hydrochloride, MedChemExpress

Product Code. 16441327
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16441327

Brand: Medchem Express HY12319A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cardiogenol C hydrochloride is a potent cell-permeable pyrimidine inducer which prompts the differentiation of ESCs into cardiomyocytes (EC50=100 nM).

Cardiogenol C hydrochloride also acts cardiomyogenic on already lineage-committed progenitor cell types with a limited degree of plasticity. Cardiogenol C hydrochloride is a useful cardiomyogenic agent and can be used as a tool to improve cardiac repair by cell transplantation therapy in animal models.

Specifications

Chemical Name or Material Cardiogenol C hydrochloride
CAS 1049741-55-0
Color White
Purity Grade Notes Research
Molecular Formula C13H17ClN4O2
Quantity 50 mg
Solubility Information DMSO : ≥ 59 mg/mL (198.82 mM) ∣H2O : 2 mg/mL (6.74 mM; Need ultrasonic)
SMILES [H]Cl.COC1=CC=C(NC2=NC=CC(NCCO)=N2)C=C1
Molecular Weight (g/mol) 296.75
Formula Weight 296.75
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.