missing translation for 'onlineSavingsMsg'
Learn More

Cardiogenol C hydrochloride, MedChemExpress

Product Code. 16485088
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485088 10 mg 10mg
16441327 50 mg 50mg
16417817 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16485088 Supplier Medchem Express Supplier No. HY12319A
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Cardiogenol C hydrochloride is a potent cell-permeable pyrimidine inducer which prompts the differentiation of ESCs into cardiomyocytes (EC50=100 nM).

Cardiogenol C hydrochloride also acts cardiomyogenic on already lineage-committed progenitor cell types with a limited degree of plasticity. Cardiogenol C hydrochloride is a useful cardiomyogenic agent and can be used as a tool to improve cardiac repair by cell transplantation therapy in animal models.

Specifications

Chemical Name or Material Cardiogenol C hydrochloride
CAS 1049741-55-0
Color White
Purity Grade Notes Research
Molecular Formula C13H17ClN4O2
Quantity 10 mg
Solubility Information DMSO : ≥ 59 mg/mL (198.82 mM) ∣H2O : 2 mg/mL (6.74 mM; Need ultrasonic)
SMILES [H]Cl.COC1=CC=C(NC2=NC=CC(NCCO)=N2)C=C1
Molecular Weight (g/mol) 296.75
Formula Weight 296.75
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.