missing translation for 'onlineSavingsMsg'
Learn More

Cardiac Troponin T Antibody (OTI8B5) [DyLight 650], Novus Biologicals™

Product Code. 30688529 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688529 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688529 Supplier Novus Biologicals Supplier No. NBP344907C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cardiac Troponin T Monoclonal antibody specifically detects Cardiac Troponin T in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cardiac Troponin T
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI8B5
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Cardiac Muscle Troponin T, Cardiomyopathy, Dilated 1D (Autosomal Dominant), Cardiomyopathy, Hypertrophic, CMD1D, CMH2, CMPD2, cTnT, LVNC6, RCM3, TnTc, troponin T type 2 (cardiac), troponin T, cardiac muscle, Troponin T2, Cardiac Type, Truncated Cardiac Troponin T
Host Species Mouse
Immunogen Native troponin complex
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7139
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.