missing translation for 'onlineSavingsMsg'
Learn More

Cardiac Troponin T Antibody (OTI5D9), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

£195.00 - £417.00

Specifications

Antigen Cardiac Troponin T
Clone OTI5D9
Dilution Western Blot 1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306302
View Documents
Novus Biologicals
NBP3-44905-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306152
View Documents
Novus Biologicals
NBP3-44905-30ul
30 μL
£195.00
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cardiac Troponin T Monoclonal antibody specifically detects Cardiac Troponin T in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications

Specifications

Cardiac Troponin T
Western Blot 1:2000
Monoclonal
Purified
RUO
PBS (pH 7.3) 1% BSA, 50% glycerol
7139
IgG2a
>95%
Antibody
OTI5D9
Western Blot
Unconjugated
Mouse
Human, Mouse, Rat
Cardiac Muscle Troponin T, Cardiomyopathy, Dilated 1D (Autosomal Dominant), Cardiomyopathy, Hypertrophic, CMD1D, CMH2, CMPD2, cTnT, LVNC6, RCM3, TnTc, troponin T type 2 (cardiac), troponin T, cardiac muscle, Troponin T2, Cardiac Type, Truncated Cardiac Troponin T
Full length human recombinant protein of human TNNT2 (NP_000355) produced in E.coli.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.