missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cardiac Troponin C Monoclonal Antibody (7B9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA122698
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-22698 detects Cardiac Troponin C in Human samples.
Troponin (Tn) is the central regulatory protein of striated muscle contraction. Tn consists of three components: Tn-I which is the inhibitor of actomyosin ATPase, Tn-T which contains the binding site for tropomyosin and Tn-C. The binding of calcium to Tn-C abolishes the inhibitory action of Tn on actin filaments, and as a result of calcium-ion uptake by troponin-C, the elements of the muscle thin filament undergo a series of allosteric changes which allow the interaction of actin with myosin, the hydrolysis of ATP, and the generation of tension.Specifications
Cardiac Troponin C | |
Monoclonal | |
6.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P63316 | |
Tnnc1 | |
Human cardiac troponin complex | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Affinity Purification, ELISA, Western Blot | |
7B9 | |
Unconjugated | |
Tnnc1 | |
AI874626; cardiac 3; cardiac troponin C; Cardiac troponin I; CMD1FF; CMD1Z; CMD2A; CMH13; CMH7; cTnC; cTnI; MGC116817; RCM1; slow twitch skeletal/cardiac muscle troponin C; Tn1; TnC; TN-C; Tncc; Tni; TNN I3; TNNC; TNNC 1; Tnnc1; TNNI3; troponin C type 1 (slow); troponin C, cardiac/slow skeletal; troponin C, slow skeletal and cardiac muscles; troponin C1, slow; troponin C1, slow skeletal and cardiac type; Troponin I; Troponin I cardiac; TroponinI | |
Mouse | |
Protein A | |
RUO | |
7134 | |
4° C, do not freeze | |
Liquid |