missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cardiac Leiomodin Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26068JF585
This item is not returnable.
View return policy
Description
Cardiac Leiomodin Polyclonal antibody specifically detects Cardiac Leiomodin in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Cardiac Leiomodin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 442721 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 585 | |
| C-LMOD, Leiomodin 2 (Cardiac), leiomodin-2, LMOD2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to Human Cardiac Leiomodin | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction