missing translation for 'onlineSavingsMsg'
Learn More

CARD9 Antibody [Biotin], Novus Biologicals™

Product Code. 30733359 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733359 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733359 Supplier Novus Biologicals Supplier No. AF5248B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CARD9 Polyclonal antibody specifically detects CARD9 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARD9
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias CANDF2, caspase recruitment domain family, member 9, caspase recruitment domain-containing protein 9, hCARD9
Host Species Sheep
Immunogen E. coli-derived recombinant human CARD9, Lys225-Gln359, Accession # Q9H257
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 64170
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.