missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARD8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA585165
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Positive Control: Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Apoptosis is related to many diseases and development. Cell death signals are transduced by death domain (DD), death effector domain (DED), and caspase recruitment domain (CARD) containing molecules. CARD containing proteins include some caspases, Apaf-1, CARD4, IAPs, RICK, ARC, RAIDD, BCL-10, and ASC. A novel CARD-containing protein was recently identified and designated CARD8. This protein interacts with DRAL, a p53-responsive protein implicated in the induction of apoptosis, and caspase-1 and its related proteins ICEBERG and pesudo-ICE. Although there are conflicting reports on whether CARD8 acts a pro- or anti-apoptotic protein, it has been suggested that it functions as an adaptor molecule regulating caspase-1 and NF-kappa-B activation.
Specifications
| CARD8 | |
| Polyclonal | |
| Unconjugated | |
| CARD8 | |
| Apoptotic protein NDPP1; CARD 8; CARD inhibitor of NF-kappaB-activating ligands; CARD8; CARDINAL; CARD-inhibitor of NF-kappa-B-activating ligand; caspase recruitment domain family member 8; caspase recruitment domain family, member 8; caspase recruitment domain-containing protein 8; DACAR; DAKAR; DKFZp779L0366; KI; KIAA0955; NDPP; NDPP1; TUCAN; Tumor up-regulated CARD-containing antagonist of CASP9; tumor up-regulated CARD-containing antagonist of caspase nine | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22900 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y2G2 | |
| CARD8 | |
| Recombinant protein encompassing a sequence within the center region of human CARD8. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction