missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CARD8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559852
This item is not returnable.
View return policy
Description
Immunogen sequence: TLVWDTEVKP VDLQLVAASA PPPFSGAAFV KENHRQLQAR MGDLKGVLDD LQDNEVLTEN EKELVEQEKT RQSKNEALLS MVEKKGDLA Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 31%.
Apoptosis is related to many diseases and development. Cell death signals are transduced by death domain (DD), death effector domain (DED), and caspase recruitment domain (CARD) containing molecules. CARD containing proteins include some caspases, Apaf-1, CARD4, IAPs, RICK, ARC, RAIDD, BCL-10, and ASC. A novel CARD-containing protein was recently identified and designated CARD8. This protein interacts with DRAL, a p53-responsive protein implicated in the induction of apoptosis, and caspase-1 and its related proteins ICEBERG and pesudo-ICE. Although there are conflicting reports on whether CARD8 acts a pro- or anti-apoptotic protein, it has been suggested that it functions as an adaptor molecule regulating caspase-1 and NF-kappa-B activation.
Specifications
| CARD8 | |
| Polyclonal | |
| Unconjugated | |
| CARD8 | |
| Apoptotic protein NDPP1; CARD 8; CARD inhibitor of NF-kappaB-activating ligands; CARD8; CARDINAL; CARD-inhibitor of NF-kappa-B-activating ligand; caspase recruitment domain family member 8; caspase recruitment domain family, member 8; caspase recruitment domain-containing protein 8; DACAR; DAKAR; DKFZp779L0366; KI; KIAA0955; NDPP; NDPP1; TUCAN; Tumor up-regulated CARD-containing antagonist of CASP9; tumor up-regulated CARD-containing antagonist of caspase nine | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22900 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2G2 | |
| CARD8 | |
| Recombinant protein corresponding to Human CARD8. Recombinant protein control fragment (Product #RP-98957). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction