missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CARD6 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518154
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a protein that contains a caspase recruitment domain (CARD), an antiparallel six-helical bundle that mediates homotypic protein-protein interactions. The encoded protein is a microtubule-associated protein that has been shown to interact with receptor-interacting protein kinases and positively modulate signal transduction pathways converging on activation of the inducible transcription factor NF-kB.
Specifications
| CARD6 | |
| Polyclonal | |
| Unconjugated | |
| CARD6 | |
| Card6; CARD-containing inhibitor of Nod1 and Cardiak-induced NF-kB activation; caspase recruitment domain family member 6; caspase recruitment domain family, member 6; caspase recruitment domain protein 6; caspase recruitment domain-containing protein 6; CINCIN1; D730008L15 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 84674 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BX69 | |
| CARD6 | |
| Synthetic peptide sequence (ATESTPSEIIERER) corresponding to the N-terminus amino acids of CARD6 (aa 2-16). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction