Learn More
Invitrogen™ CARD6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589451
Description
Immunogen sequence: MATESTPSEI IERERKKLLE ILQHDPDSIL DTLTSRRLIS EEEYETLENV TDLLKKSRKL LILVQKKGEA TCQHFLKCLF STFPQSAAIC GLRHEVLKHE NTVPPQSMGA SSNSEDAFSP GIKQPEAPEI TVFFSEKEHL DLETSEFFRD KKTSYRETAL SARKNEKEYD TPEVTLSYSV EKVGCEVPAT ITYIKDGQRY; Positive Samples: HT-29, MCF-7, 293T, HepG2.
This gene encodes a protein that contains a caspase recruitment domain (CARD), an antiparallel six-helical bundle that mediates homotypic protein-protein interactions. The encoded protein is a microtubule-associated protein that has been shown to interact with receptor-interacting protein kinases and positively modulate signal transduction pathways converging on activation of the inducible transcription factor NF-kB.
Specifications
| CARD6 | |
| Polyclonal | |
| Unconjugated | |
| CARD6 | |
| Card6; CARD-containing inhibitor of Nod1 and Cardiak-induced NF-kB activation; caspase recruitment domain family member 6; caspase recruitment domain family, member 6; caspase recruitment domain protein 6; caspase recruitment domain-containing protein 6; CINCIN1; D730008L15 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84674 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.44 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9BX69 | |
| CARD6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human CARD6 (NP_1159762). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.