missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARD6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92129-0.02ml
This item is not returnable.
View return policy
Description
CARD6 Polyclonal antibody specifically detects CARD6 in Human samples. It is validated for Western Blot
Specifications
| CARD6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| caspase recruitment domain family, member 6, caspase recruitment domain protein 6, caspase recruitment domain-containing protein 6, CINCIN1CARD-containing inhibitor of Nod1 and Cardiak-induced NF-kB activation | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human CARD6 (NP_115976.2). MATESTPSEIIERERKKLLEILQHDPDSILDTLTSRRLISEEEYETLENVTDLLKKSRKLLILVQKKGEATCQHFLKCLFSTFPQSAAICGLRHEVLKHENTVPPQSMGASSNSEDAFSPGIKQPEAPEITVFFSEKEHLDLETSEFFRDKKTSYRETALSARKNEKEYDTPEVTLSYSVEKVGCEVPATITYIKDGQRY | |
| 0.02 mL | |
| Apoptosis | |
| 84674 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction