missing translation for 'onlineSavingsMsg'
Learn More

CARD14 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18418436 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418436 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418436 Supplier Novus Biologicals Supplier No. NBP281901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CARD14 Polyclonal antibody specifically detects CARD14 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARD14
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias bcl10-interacting maguk protein 2, BIMP2, CARD-containing MAGUK protein 2, card-maguk protein 2, Carma 2, CARMA2CARD-containing MAGUK 2 protein, caspase recruitment domain family, member 14, caspase recruitment domain-containing protein 14
Host Species Rabbit
Immunogen CARMA2 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human CARMA2. The immunogen is located within the last 50 amino acids of CARMA2. Amino Acid Squence: DEQKKVVWTEQSPR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 79092
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.