missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARD12 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£169.00 - £400.00
Specifications
| Antigen | CARD12 |
|---|---|
| Concentration | 1.13 mg/mL |
| Dilution | Flow Cytometry reported in scientific literature (PMID 29104144), Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:400, Immunoblotting reported in scientific literature (PMID 29104144) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18229825
|
Novus Biologicals
NBP1-78979SS |
0.025 mL |
£169.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18216638
|
Novus Biologicals
NBP1-78979 |
0.1 mL |
£400.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CARD12 Polyclonal specifically detects CARD12 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
| CARD12 | |
| Flow Cytometry reported in scientific literature (PMID 29104144), Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:400, Immunoblotting reported in scientific literature (PMID 29104144) | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| CARD12leucine rich repeat and CARD domaincontaining 4, caspase recruitment domain family, member 12, Caspase recruitment domain-containing protein 12, Clan protein, CLAN1ipaf, CLANCARD, LRR, and NACHT-containing protein, CLR2.1, Ice protease-activating factor, Ipaf, NLR family, CARD domain containing 4 | |
| NLRC4 | |
| IgG | |
| Affinity Purified |
| 1.13 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q9NPP4 | |
| 58484 | |
| A synthetic peptide made to an internal portion of the human CARD12 protein (between residues 600-700) [UniProt Q9NPP4] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title