missing translation for 'onlineSavingsMsg'
Learn More

CARD11/CARMA1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18320227 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320227 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320227 Supplier Novus Biologicals Supplier No. NBP299265JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CARD11/CARMA1 Polyclonal specifically detects CARD11/CARMA1 in Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARD11/CARMA1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50 mM Sodium Borate
Gene Alias BIMP3card-maguk protein 1, CARD-containing MAGUK protein 1, Carma 1, CARMA1bcl10-interacting maguk protein 3, caspase recruitment domain family, member 11, caspase recruitment domain-containing protein 11, MGC133069
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse CARD11/CARMA1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 84433
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.