missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CARD10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534364
This item is not returnable.
View return policy
Description
A suggested positive control is human kidney tissue. PA5-34364 can be used with blocking peptide PEP-1407.
CARMA proteins belong to the membrane-associated guanylate kinase-like (MAGUK) family of proteins that can function as molecular scaffolds that assist assembly of signal transduction molecules. CARMA1, CARMA2, and CARMA3 share high degrees of sequence and functional homology, but their tissue-specific distribution suggests that they serve distinct biological functions in different cell types. As with CARMA1, the CARD domain of CARMA3 has been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-Kappa-B activation. When expressed in cells, this protein binds to BCL10 and activates NF-Kappa-B Recent experiments have shown that CARMA3 is required for EGF-induced NF-Kappa-B activation and contributes to tumor growth in vivo, suggesting that CARMA3 may serve as a new therapeutic target for the treatment of EGFR-driven tumors.
Specifications
| CARD10 | |
| Polyclonal | |
| Unconjugated | |
| CARD10 | |
| AI449026; Bcl10 binding protein and activator of NFKB; bcl10-interacting MAGUK protein 1; Bimp1; Card10; CARD-containing MAGUK 3 protein; CARD-containing MAGUK protein 3; Carma 3; Carma3; caspase recruitment domain family member 10; caspase recruitment domain family, member 10; caspase recruitment domain-containing protein 10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29775 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9BWT7 | |
| CARD10 | |
| A 16 amino acid synthetic peptide near the amino terminus of human CARMA3. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction