missing translation for 'onlineSavingsMsg'
Learn More

CARD10 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18424248 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424248 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424248 Supplier Novus Biologicals Supplier No. NBP176676FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CARD10 Polyclonal antibody specifically detects CARD10 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARD10
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CARD-containing MAGUK 3 protein, CARD-containing MAGUK protein 3, Carma 3, CARMA3BIMP1Bcl10 binding protein and activator of NFKB, caspase recruitment domain family, member 10, caspase recruitment domain-containing protein 10, MGC142219
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 14 amino acids at the C-terminus of human CARD10. The immunogen is located within the last 50 amino acids of CARD10. Amino Acid Squence: KECGSSRGCPSSSED
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 29775
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.