missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Carcinoembryonic Antigen (CEA)

Product Code. 16402749
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402749 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402749 Supplier MedChemExpress Supplier No. HYP0277
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Carcinoembryonic Antigen (CEA) is a tumor marker in lung cancer.

Specifications

Purity 99.49%
CAS 168635-85-6
Content And Storage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 964.07 g/mol
Product Type Natural Products
Quantity 1 mg
Shipping Condition Room temperature
Solubility H2O : 12.5 mg/mL (12.97 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.